

1,1-Dibromoethane 99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:557-91-5
:
Formula:Br₂HCCH₃
MDL Number:MFCD00041719
:187.86 g/mol
Boiling Pt:107 °C
Melting Pt:–0.5 °C
Density:2.06 g/cm³
Storage Temperature:Ambient
EINECS:209-184-9
UN:2810
ADR:6.1,II
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear colorless to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5090 to 1.5120 (20°C, 589 nm)
- GC:≥98.5 %
- Stabilizer:potassium carbonate