Tin(II) chloride dihydrate 98+%
:10025-69-1
:
:SnCl₂·2H₂O
:MFCD00149863
:225.65 g/mol
:13,08861
:652 °C (760 mmHg)
:37…38 °C
:2,71 g/cm³ (25 °C)
:Ambient
:231-868-0
:3260
:8,III
:GHS07
:GHS08
:GHS05
- Appearance:Colorless to white Crystalline powder or crystals and/or chunks
- Titration Iodimetric:98.0 to 103.0 %