Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminium nitrate nonahydrate
Aluminium nitrate nonahydrate ≥98%
Aluminium nitrate nonahydrate ≥98%
:
Thermo Fisher Scientific
Aluminium nitrate nonahydrate ≥98%
:
Thermo Fisher Scientific
:
7784-27-2
:
:
Al(NO₃)₃·9H₂O
:
MFCD00149132
:
375.13 g/mol
:
14,352
:
73.5 °C
:
1.72 g/cm³ (20 °C)
:
Ambient
:
236-751-8
:
1438
:
5.1,III
:
GHS03
:
GHS07
msds_ALFA012360
Thermo Scientific Acroseal 25 Years