Order Entry
Export
ContactUsLinkComponent
Aluminium nitrate nonahydrate 99+% for analysis
Corrosive
Irritant
Aluminium nitrate nonahydrate 99+% for analysis
Certificates
undefined
Corrosive
Irritant
Aluminium nitrate nonahydrate 99+% for analysis
Certificates

Specification Test Results

  • Appearance:
    Semitransparent to transparent Crystals
  • Titration Complexometric:
    ≥99.0 %
  • pH:
    2.5 to 3.5 (5 % at 20°C)
  • Chloride (Cl):
    ≤20 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Iron (Fe):
    ≤20 ppm
  • Heavy metals (as Pb):
    ≤20 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number