Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Diiodopropane
1,3-Diiodopropane ≥98% stabilised
1,3-Diiodopropane ≥98% stabilised
:
Thermo Fisher Scientific
1,3-Diiodopropane ≥98% stabilised
:
Thermo Fisher Scientific
:
627-31-6
:
:
C₃H₆I₂
:
MFCD00001097
:
295.89 g/mol
:
224 °C
:
–20 °C
:
2.565
:
>110 °C (230 °F)
:
Stabilised with copper
:
210-993-4
:
GHS07
:
Ambient
msds_ALFAA10616
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years