
Calcium chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7774-34-7
:
Formula:CaCl₂·6H₂O
MDL Number:MFCD00149616
:219.08 g/mol
Merck Index:13,01660
Melting Pt:29.54 °C
Density:1.71 g/cm³ (25 °C)
EINECS:233-140-8
:GHS07
Storage Temperature:Ambient