Order Entry
Export
ContactUsLinkComponent
 
Citric acid monohydrate
Catalog # ROTH1818.1
Supplier:  Roth Carl
CAS Number:  
undefined
Citric acid monohydrate
Catalog # ROTH1818.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications

About this item

 :
 :210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1.552 g/cm³ (20 °C)
Flash Pt:173.9 °C (closed cup)
EINECS:201-069-1
 :GHS07
Storage Temperature:Ambient
CAS Number:5949-29-1
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972