Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(+)-Camphor
D-(+)-Camphor ≥98%
D-(+)-Camphor ≥98%
:
Thermo Fisher Scientific
D-(+)-Camphor ≥98%
:
Thermo Fisher Scientific
:
464-49-3
:
:
C₁₀H₁₆O
:
MFCD00064149
:
152.24 g/mol
:
14,1732
:
204 °C
:
175…180 °C
:
0.990
:
65 °C (149 °F)
:
Ambient
:
207-355-2
:
2717
:
4.1,III
:
GHS02
:
GHS07
msds_ALFAA10708
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years