Formic acid 1 mol/l, Sigma-Aldrich®
Catalog # SIAL06473-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:46.03 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-579-1
UN:3412
REACH:01-2119491174-37-XXXX
CAS Number:64-18-6
Formula:HCO₂H
MDL Number:MFCD00003297