(±)-3-Methylcyclohexanone
Catalog # EHERC15085020
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:112.17 g/mol
Boiling Pt:169 °C (1013 hPa)
Melting Pt:–73 °C
Density:0.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-710-7
UN:1993
ADR:3,III
CAS Number:591-24-2
Formula:C₇H₁₂O
MDL Number:MFCD00001639