

(±)-3-Methylcyclohexanone 97%
:591-24-2
:
:C₇H₁₂O
:MFCD00001639
:112.17 g/mol
:169 °C (1013 hPa)
:–73 °C
:0.915 g/cm³ (20 °C)
:48 °C
:Ambient
:209-710-7
:2297
:3,III
:GHS07
:GHS02
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4440 to 1.4460 (20°C, 589 nm)
- GC:≥96.0 %