0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Hydrogen peroxide
Hydrogen peroxide 35%
Hydrogen peroxide 35% 35%
Plastic bottle for solids, 500 g
WAKO634-03365
:
Wako
Hydrogen peroxide 35% 35%
WAKO634-03365
:
Wako
:
7722-84-1
Hydrogen peroxide 35% 35%
Plastic bottle for solids, 500 g
:
7722-84-1
599,00 €
1 * 500 g
-
+
:
7722-84-1
:
H₂O₂
:
MFCD00011333
:
34.01 g/mol
:
∼107 °C (1013 hPa)
:
< 0 °C
:
1.11 g/cm³ (20 °C)
:
Ambient
:
231-765-0
:
2014
:
5.1,II
:
Density: 1.11 g/cm³ (20 °C)
Pack type:
Plastic bottle for solids
Pk:
500 g