Order Entry
Export
Orders LinkContactUsLinkComponent
 

Nitric acid 60%

Plastic bottle, 250 mL
Catalog # 29698.233
Corrosive
Supplier:   VWR Chemicals
Nitric acid 60%
Catalog # 29698.233
Supplier:  VWR Chemicals
CAS Number:  7697-37-2
Nitric acid 60%
Nitric acid 60%

Plastic bottle, 250 mL

CAS Number:  7697-37-2
Price
Inquire For Price
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  Nitric acid solution
  • CAS Number:7697-37-2
  • Formula:HNO₃
  • MDL Number:MFCD00011349
  • Molecular Weight:63.01 g/mol
  • Boiling Pt:∼120 °C (1013 hPa)
  • Melting Pt:–22 °C
  • Density:1.37 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-714-2
  • UN:2031
  • ADR:8,II
  •  :Density: 1.51 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL

Specifications