Order Entry
Export
Orders LinkContactUsLinkComponent
Hydrochloric acid 20%

Hydrochloric acid 20%

Glass bottle for solids, 500 g
Catalog # WAKO639-03315
Irritant
Supplier:   Wako
Hydrochloric acid 20%
Catalog # WAKO639-03315
Supplier:  Wako
CAS Number:  7647-01-0
Hydrochloric acid 20%

Glass bottle for solids, 500 g

CAS Number:  7647-01-0
Price
599,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications