Formic acid 90%
Plastic container, 5 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Aminic Acid, Methanoic acid
- CAS Number:64-18-6
- Formula:HCO₂H
- MDL Number:MFCD00003297
- Molecular Weight:46.03 g/mol
- Storage Temperature:Ambient
- ADR:8,II
- :Density: 1.22 g/cm³ (25 °C)
- EINECS:200-579-1
- UN:1779
Documents
Specifications
Specifications
- Pack type:Plastic container
- Pk:5 L



