



1,3-Dichloropropene
Catalog # EHERC12490000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:110.97 g/mol
Merck Index:13,03101
Boiling Pt:108 °C (1013 hPa)
Melting Pt:–83,9 °C
Density:1,25 g/cm³ (0 °C)
Storage Temperature:Ambient
EINECS:208-826-5
UN:2047
ADR:3,II
:GHS02
:GHS06
:GHS08
:GHS09
CAS Number:542-75-6
Formula:C₃H₄Cl₂
MDL Number:MFCD00000985