Order Entry
Export
ContactUsLinkComponent
1,2-Dibromoethylene
1,2-Dibromoethylene
Catalog # EHERCA12240200
Supplier:  EHRENSTORFER
CAS Number:  
1,2-Dibromoethylene
Catalog # EHERCA12240200
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :185.85 g/mol
Merck Index:12,00092
Boiling Pt:111…112 °C (1013 hPa)
Melting Pt:–53 °C
Density:2.268 g/cm³ (0 °C)
EINECS:208-747-6
UN:2810
ADR:6.1,III
CAS Number:540-49-8
Formula:C₂H₂Br₂
MDL Number:MFCD00000184