Zinc, powder
Supplier: Roth Carl
Jump to:
About this item
- CAS Number:7440-66-6
- Formula:Zn
- MDL Number:MFCD00011291
- Molecular Weight:65.39 g/mol
- Boiling Pt:907 °C (1013 hPa)
- Melting Pt:419.5 °C
- Density:7.13 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-175-3
- UN:1436
- ADR:4.3,II
2 Options Available Below
Product Details & Documents
- CAS Number:7440-66-6
- Formula:Zn
- MDL Number:MFCD00011291
- Molecular Weight:65.39 g/mol
- Boiling Pt:907 °C (1013 hPa)
- Melting Pt:419.5 °C
- Density:7.13 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-175-3
- UN:1436
- ADR:4.3,II
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



