About this item
PS, optically clear. BRANDplates® 96-well strip plates have ideal optical characteristics and are manufactured according to ANSI and SLAS standards. They offer high flexibility as only as many wells as necessary need be used. To fit any application, strip plates are available without or with grid, in versions with high-binding capacity or medium-binding capacity.
immunoGrade™: Surface with high-binding capacity, optimised for the immobilisation of IgG and molecules with hydrophobic and hydrophilic regions.
pureGrade™: Non treated surface with medium-binding capacity, somewhat more hydrophobic than immunoGrade™.
Strip plates without grid: 12 strips of 8 wells with flat bottom. Individual strips can be removed from the frame and reinserted. The strips are not divisible.
Strip plates with grid: 12 strips of 8 wells with flat bottom, divisible. Individual strips are removable from the plate and can be reinserted. The strips can be divided into 8 individual vessels and can be inserted individually into the frame plate.