Order Entry
Export
ContactUsLinkComponent
Zinc ≥99.9998% (metals basis), rods, Ø 10 mm (0.39 in)
Zinc ≥99.9998% (metals basis), rods, Ø 10 mm (0.39 in)
Catalog # 12718.DT
CAS Number:  
Zinc ≥99.9998% (metals basis), rods, Ø 10 mm (0.39 in)
Catalog # 12718.DT
CAS Number:  

Specifications

  • Pk:
    250 mm
  • Seq:
    1856

Specifications

About this item

 :
 :65.39 g/mol
Merck Index:12,10255
Boiling Pt:908 °C (1013 hPa)
Melting Pt:420 °C
Density:7.14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-175-3
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291