

Zinc, granular 30 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-66-6
:
Formula:Zn
MDL Number:MFCD00011291
:65.39 g/mol
Merck Index:12,10255
Boiling Pt:908 °C (1013 hPa)
Melting Pt:420 °C
Density:7,14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-175-3
UN:1436
:GHS02
:GHS09
Specification Test Results
- Appearance:Silver grey Granules or filings
- Titration Complexometric:≥99.8 %
- Particle size:30 mesh
- Suitability:(For determination of As) Passes test
- Iron (Fe):≤0.01 %
- Lead (Pb):≤0.01 %