Order Entry
Export
ContactUsLinkComponent
Copper ≥99.999% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)
Copper ≥99.999% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)
Catalog # 11377.FI
CAS Number:  
Copper ≥99.999% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)
Catalog # 11377.FI
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Pk Info:
    50×50 mm

Specifications

About this item

 :
 :63.55 g/mol
Merck Index:13,02545
Boiling Pt:2595 °C (1013 hPa)
Melting Pt:1083 °C
Density:8.94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-159-6
CAS Number:7440-50-8
Formula:Cu
MDL Number:MFCD00010965