Copper 99%, powder max 106 µm (-140 mesh)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-50-8
:
Formula:Cu
MDL Number:MFCD00010965
:63.55 g/mol
Merck Index:13,02545
Boiling Pt:2595 °C (1013 hPa)
Melting Pt:1083 °C
Density:8,94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-159-6
UN:3089
ADR:4.1,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Red to red-brown Powder
- Assay:≥98.5 % (Cu)
- Particle size:(Maximum) 106 µm
- Apparent density:1.35 g/ml
- Average particle size:25 µm