
Tellurium 99+% (trace metals basis), powder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13494-80-9
:
Formula:Te
MDL Number:MFCD00134062
:127.6 g/mol
Merck Index:12,09273
Boiling Pt:990 °C (1013 hPa)
Melting Pt:450 °C
Density:8,83 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-813-4
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Grey to black Powder
- Total trace metal impurities:≤1.0 %
- Melting point:77 to 81 °C
- GC:≥97.5 %