
Tellurium 99.8% (trace metals basis), powder -200 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13494-80-9
:
Formula:Te
MDL Number:MFCD00134062
:127.6 g/mol
Merck Index:12,09273
Boiling Pt:990 °C (760 mmHg)
Melting Pt:450 °C
Density:8,83 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-813-4
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Grey to black Powder
- Total trace metal impurities:≤0.2 %