Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antimony
Antimony ≥99.999% (metals basis), powder -200 mesh
Antimony ≥99.999% (metals basis), powder -200 mesh
:
Thermo Fisher Scientific
Antimony ≥99.999% (metals basis), powder -200 mesh
:
Thermo Fisher Scientific
:
7440-36-0
:
:
Sb
:
MFCD00134030
:
121.76 g/mol
:
Ambient
:
231-146-5
:
2871
:
6.1,III
:
GHS06
:
GHS09
msds_ALFA014640
Thermo Scientific Acroseal 25 Years