

Antimony ≥99.99% (metals basis), ingot
Catalog # 00030.A1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
CAS Number:7440-36-0
:
Formula:Sb
MDL Number:MFCD00134030
:121.76 g/mol
Merck Index:12,00733
Boiling Pt:1637 °C (1013 hPa)
Melting Pt:630 °C
Density:6.684 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-146-5
UN:2871
ADR:6.1,III
:GHS07
:GHS09