Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lead
Lead ≥99.999% (metals basis), granules 0.6-4.0 mm, (0.02-0.12 in)
Lead ≥99.999% (metals basis), granules 0.6-4.0 mm, (0.02-0.12 in)
:
Thermo Fisher Scientific
Lead ≥99.999% (metals basis), granules 0.6-4.0 mm, (0.02-0.12 in)
:
Thermo Fisher Scientific
:
7439-92-1
:
:
Pb
:
MFCD00134050
:
207.2 g/mol
:
12,05410
:
1740 °C (1013 hPa)
:
327 °C
:
11.3437 g/cm³ (20 °C)
:
231-100-4
:
GHS08
:
GHS07
:
GHS09
:
Ambient
externalurl.pdf
msds_ALFA041370
Thermo Scientific Acroseal 25 Years