Order Entry
Export
ContactUsLinkComponent
Lead ≥99.9% (metals basis), wire, Ø 1.0 mm (0.04 in)
Health
Irritant
Environment
Lead ≥99.9% (metals basis), wire, Ø 1.0 mm (0.04 in)
Catalog # 12605.G9
CAS Number:  
Health
Irritant
Environment
Lead ≥99.9% (metals basis), wire, Ø 1.0 mm (0.04 in)
Catalog # 12605.G9
CAS Number:  

Specifications

  • Pk:
    25 m
  • Seq:
    8406

Specifications

About this item

 :
 :207.2 g/mol
Merck Index:12,05410
Boiling Pt:1740 °C (1013 hPa)
Melting Pt:327 °C
Density:11.3437 g/cm³ (20 °C)
EINECS:231-100-4
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:7439-92-1
Formula:Pb
MDL Number:MFCD00134050