Order Entry
Export
ContactUsLinkComponent
Sodium 99%, sticks, coated in film of protective hydrocarbon oil
Flammable
Health
Corrosive
Sodium 99%, sticks, coated in film of protective hydrocarbon oil
Catalog # L13285.36
CAS Number:  
Flammable
Health
Corrosive
Sodium 99%, sticks, coated in film of protective hydrocarbon oil
Catalog # L13285.36
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :22.99 g/mol
Merck Index:14,8570
Boiling Pt:881 °C
Melting Pt:98 °C
Density:0.9 g/cm³ (20 °C)
Flash Pt:>110 °C (230 °F)
EINECS:231-132-9
UN:1428
ADR:4.3,I
 :GHS02
 :GHS08
 :GHS05
Storage Temperature:Ambient
CAS Number:7440-23-5
Formula:Na
MDL Number:MFCD00085307