ContactUsLinkComponent
Sterilisation wraps, Bio-Shield®
  DUPO4072
 :  DuPont
undefined
Sterilisation wraps, Bio-Shield®
  DUPO4072
 :  DuPont
  • Colour:
    Blue
  • Description:
    Regular sterilisation wrap
  • Dimensions:
    1828,8×1371,6 mm
  • Seq:
    19
  • Pk:
    100
These versatile wraps offer fluid repellency and provide an excellent barrier against airborne bacteria. The soft, drapable non woven blend of cellulose and polyester is virtually lint-free and helps protect wrapped contents from contamination.
  • For light trays, small basins and towels
  • Sterilisation tapes adhere firmly and remove easily
  • Superior bacterial barrier properties compared to linen
  • Water and tear resistant