Capsaicin, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:404-86-4
:
Formula:C₁₈H₂₇NO₃
MDL Number:MFCD00017259
:305.42 g/mol
Merck Index:12,01811
Boiling Pt:210…220 °C (0.01 mmHg)
Melting Pt:66.1 °C
EINECS:206-969-8
UN:2811
ADR:6.1,III