Order Entry
Export
Orders LinkContactUsLinkComponent
 

Capsaicin

Capsaicin
Supplier:  Cayman Chemical
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  8-Methyl-N-vanillyl-6-none-amid
  • CAS Number:404-86-4
  • Formula:C₁₈H₂₇NO₃
  • MDL Number:MFCD00017259
  • Molecular Weight:305.42 g/mol
  • Merck Index:12,01811
  • Boiling Pt:210…220 °C (0.01 mmHg)
  • Melting Pt:66.1 °C
  • EINECS:206-969-8
  • UN:2811
  • ADR:6.1,III

 

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 46,00 €
    46,00 €
    1 * 500 mg
       46,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 87,00 €
    87,00 €
    1 * 1 g
       87,00 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 302,00 €
    302,00 €
    1 * 5 g
       302,00 €