Order Entry
Export
ContactUsLinkComponent
Sodium selenate decahydrate ≥97.0%, GPR RECTAPUR®
Toxic
Health
Environment
Sodium selenate decahydrate ≥97.0%, GPR RECTAPUR®
Catalog # 302113L
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Health
Environment
Sodium selenate decahydrate ≥97.0%, GPR RECTAPUR®
Catalog # 302113L
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    Min. 97.0 %
  • Metal impurities:
    Max. 0.02 %
  • Insolubility in water:
    Max. 0.05 %

Specification Test Results

About this item

 :
Melting Pt:35 °C
Density:1.584 g/cm³ (20 °C)
UN:2630
ADR:6.1,I
 :369.09 g/mol
EINECS:236-501-8
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:10102-23-5
Formula:Na₂SeO₄·10H₂O
MDL Number:MFCD00149174