
Xylene cyanole, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4463-44-9
:
Formula:C₂₅H₂₇N₂NaO₇S₂
MDL Number:MFCD00019481
:554.61 g/mol
EINECS:224-728-5
:GHS07
Specification Test Results
- Appearance:Dark red to dark blue Crystalline powder
- Infrared spectrum:Conforms
- Dye content:≥80 %
- GC:≥99.4 %