4,4'-Dimethylbenzophenone 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:611-97-2
:
Formula:C₁₅H₁₄O
MDL Number:MFCD00017214
:210.28 g/mol
Boiling Pt:200 °C (17 mmHg)
Melting Pt:93…97 °C
Density:1,18 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-287-6
Specification Test Results
- Appearance:White to light brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:93 to 97 °C
- GC:≥98.0 %