Order Entry
Export
Orders LinkContactUsLinkComponent
4-Bromo-3-chlorophenol ≥98%

4-Bromo-3-chlorophenol ≥98%

Glass bottle for solids, 5 g
Catalog # L13801.06
Irritant
4-Bromo-3-chlorophenol ≥98%
Catalog # L13801.06
CAS Number:  13631-21-5
4-Bromo-3-chlorophenol ≥98%
4-Bromo-3-chlorophenol ≥98%

Glass bottle for solids, 5 g

CAS Number:  13631-21-5
Price
112,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98% CAS No.: 13631-21-5 Molecular Formula: C6H4BrClO Formula Weight: 207.46 Odour: Phenol-like Melting Point: 64-68° Beilstein Reference: 3236445 MDL No.: MFCD00070744

  • CAS Number:13631-21-5
  • Formula:C₆H₄BrClO
  • MDL Number:MFCD00070744
  • Molecular Weight:207.45 g/mol
  • Melting Pt:64…68 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications