Order Entry
Export
ContactUsLinkComponent
2,2',4,4'-Tetrahydroxybenzophenone
2,2',4,4'-Tetrahydroxybenzophenone
Catalog # MOLE11590326-100G
Supplier:  Molekula
CAS Number:  
2,2',4,4'-Tetrahydroxybenzophenone
Catalog # MOLE11590326-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :246.22 g/mol
Boiling Pt:100…102 °C (40 mmHg)
Melting Pt:196…198 °C
Density:1,498 g/cm³ (20 °C)
EINECS:205-028-9
CAS Number:131-55-5
Formula:C₁₃H₁₀O₅
MDL Number:MFCD00002278