

α,α,α',α'-Tetrabromo-o-xylene, Sigma-Aldrich®
Catalog # SIALT5605-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:421.75 g/mol
Boiling Pt:346 °C (1013 hPa)
Melting Pt:117 °C
ADR:6.1,II
:M-Clarity™ quality level = MQ100
EINECS:236-176-2
UN:3143
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:13209-15-9
Formula:C₈H₆Br₄
MDL Number:MFCD00000131