Order Entry
Export
ContactUsLinkComponent
Zinc carbonate basic 58% (53% Zn min.), powder, extra pure
Irritant
Environment
Zinc carbonate basic 58% (53% Zn min.), powder, extra pure
Catalog # ACRO222650010
CAS Number:  
undefined
Irritant
Environment
Zinc carbonate basic 58% (53% Zn min.), powder, extra pure
Catalog # ACRO222650010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Titration Complexometric:
    ≥53 % (Zn)
  • Chloride (Cl):
    ≤0.3 %
  • Sulfate (SO4):
    ≤1.5 %

Specification Test Results

About this item

 :
 :549.01 g/mol
Melting Pt:> 140 °C
Density:4.39 g/cm³
EINECS:226-076-7
UN:3077
 :GHS07
 :GHS09
CAS Number:5263-02-5
 :51839-25-9
Formula:C₂H₆O₁₂Zn₅
MDL Number:MFCD00011519