ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt (NADPH-Na4, reduced form)
Catalog # ROCH10107824001
Supplier: ROCHE
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:833.35 g/mol
:M-Clarity™ quality level = MQ100
EINECS:220-163-3
CAS Number:2646-71-1
Formula:C₂₁H₂₆N₇Na₄O₁₇P₃
MDL Number:MFCD00036263