Order Entry
Export
ContactUsLinkComponent
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form)
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form)
Catalog # ROCH10128023001
Supplier:  ROCHE
CAS Number:  
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form)
Catalog # ROCH10128023001
Supplier:  ROCHE
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :709.41 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:210-123-3
CAS Number:606-68-8
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂
MDL Number:MFCD00036200