Order Entry
Export
ContactUsLinkComponent
Haemalum (Mayer's)
Irritant
Haemalum (Mayer's)
Catalog # CTEH640400
Supplier:  CLIN TECH
CAS Number:  
Irritant
Haemalum (Mayer's)
Catalog # CTEH640400
Supplier:  CLIN TECH
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
Density:1.05 g/cm³ (20 °C)
 :GHS07