
Fast green FCF, pure, certified
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2353-45-9
:
Formula:C₃₇H₃₄N₂Na₂O₁₀S₃
MDL Number:MFCD00013053
:808.86 g/mol
Merck Index:13,03970
Melting Pt:290 °C (dec.)
EINECS:219-091-5
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Bordeaux to brown Powder
- Infrared spectrum:Conforms
- Dye content:≥85 %
- UV Lambda max:622 to 626 nm
- Absorption Ratio P-15/P+15:0.98 to 1.20
- BSC Certification:Passes