Anti-HSA Rabbit Polyclonal Antibody
Catalog # FITZ20-AR05
Supplier: FITZGERALD INDUSTRIES INTL
Specifications
- Antigen name:Human Serum Albumin
- Antigen symbol:HSA
- Clonality:Polyclonal
- Host:Rabbit
- Storage buffer:PBS with 0,05% NaN3
- Storage temperature:Store at 4°C for short term storage. Aliquot and store at –20 °C for long term storage. Avoid repeated freeze/thaw cycles.
- Immunogen:Human serum albumin antibody was raised in rabbit using human albumin as the immunogen
- Purification:Chromatographically purified
- Pack type:Glass bottle
- Pk:1 mL
Specifications
About this item
Human serum albumin is the most abundant protein in human blood plasma. It is produced in the liver. Albumin comprises about half of the blood serum protein. It is soluble and monomeric. The gene for albumin is located on chromosome 4 and mutations in this gene can result in various anomalous proteins. The human albumin gene is 16,961 nucleotides long from the putative ?cap? site to the first poly(A) addition site. It is split into 15 exons that are symmetrically placed within the 3 domains thought to have arisen by triplication of a single primordial domain.