Order Entry
Export
Orders LinkContactUsLinkComponent
Orange G

Orange G

Glass bottle for solids, 25 g
Catalog # MOLE42617597-25G
Supplier:   Molekula
Orange G
Catalog # MOLE42617597-25G
Supplier:  Molekula
CAS Number:  1936-15-8
Orange G

Glass bottle for solids, 25 g

CAS Number:  1936-15-8
Price
105,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate, Wool Orange 2G, Orange G sodium salt, CI 16230, Acid Orange 10
  • CAS Number:1936-15-8
  • Formula:C₁₆H₁₀N₂Na₂O₇S₂
  • MDL Number:MFCD00012457
  • Molecular Weight:452.38 g/mol
  • Melting Pt:141 °C
  • Density:0.45…0.47 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:217-705-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications