Order Entry
Export
ContactUsLinkComponent
2,7-Dichlorofluorene ≥97%
Irritant
2,7-Dichlorofluorene ≥97%
Catalog # B24717.06
CAS Number:  
Irritant
2,7-Dichlorofluorene ≥97%
Catalog # B24717.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :235.11 g/mol
Melting Pt:>300 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:7012-16-0
Formula:C₁₃H₈Cl₂
MDL Number:MFCD00032840