Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ω-Chloro-3,4-dihydroxyacetophenone
ω-Chloro-3,4-dihydroxyacetophenone, Sigma-Aldrich®
ω-Chloro-3,4-dihydroxyacetophenone, Sigma-Aldrich®
SIALC34405-25G
:
Merck
:
ω-Chloro-3,4-dihydroxyacetophenone, Sigma-Aldrich®
SIALC34405-25G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
186.59
:
M-Clarity™ quality level = MQ100
:
202-754-8
:
GHS07
:
99-40-1
:
C₈H₇ClO₃
:
MFCD00002200