

(-)-Quinine hydrochloride dihydrate, Sigma-Aldrich®
Catalog # SIAL22632-50G-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:396.91 g/mol
Melting Pt:158…160 °C
:M-Clarity™ quality level = MQ100
Flash Pt:122 °C
EINECS:205-001-1
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:6119-47-7
Formula:C₂₀H₂₅ClN₂O₂·2H₂O
MDL Number:MFCD00151248