

(-)-Quinine hydrochloride dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6119-47-7
:
Formula:C₂₀H₂₅ClN₂O₂·2H₂O
MDL Number:MFCD00151248
:396.91 g/mol
Melting Pt:158…160 °C
Flash Pt:122 °C
EINECS:205-001-1
:GHS08
:GHS07
Storage Temperature:Ambient